siRNA Details
| Gene Symbol: Gm960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033447.3 | NP_001028619.2 | 2 | 541 | ||
| GenBank mRNA | AK134945.1 | - | 2 | 189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm960| Gene Symbol: Gm960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033447.3 | NP_001028619.2 | 2 | 541 | ||
| GenBank mRNA | AK134945.1 | - | 2 | 189 | ||