siRNA Details
Gene Symbol: Fam189a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114174.1 | NP_001107646.1 | 10 | 1427 | ||
GenBank mRNA | BC099492.1 | - | 5 | 551 | ||
DV649334.1 | - | 5 | 522 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam189a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114174.1 | NP_001107646.1 | 10 | 1427 | ||
GenBank mRNA | BC099492.1 | - | 5 | 551 | ||
DV649334.1 | - | 5 | 522 |