siRNA Details
| Gene Symbol: March4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045533.1 | NP_001038998.1 | 1 | 1276 | ||
| GenBank mRNA | AK122502.1 | - | 1 | 1642 | ||
| BC098191.1 | - | 1 | 1276 | 6832500 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

March4| Gene Symbol: March4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001045533.1 | NP_001038998.1 | 1 | 1276 | ||
| GenBank mRNA | AK122502.1 | - | 1 | 1642 | ||
| BC098191.1 | - | 1 | 1276 | 6832500 | ||