siRNA Details
| Gene Symbol: Mnda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033450.4 | NP_001028622.1 | 4, 5 | 866 | ||
| NM_001301745.1 | NP_001288674.1 | 4, 5 | 860 | |||
| GenBank mRNA | AK152278.1 | - | 4, 5 | 734 | ||
| AK152621.1 | - | 400 | ||||
| AK152683.1 | - | 4, 5 | 734 | |||
| AK152786.1 | - | 4, 5 | 743 | |||
| AK166217.1 | - | 4, 5 | 866 | |||
| BC132314.1 | - | 4, 5 | 687 | 40130591 | ||
| BC132316.1 | - | 4, 5 | 687 | 40130593 | ||
| BC145397.1 | - | 4, 5 | 681 | 9053955 | ||
| M74123.1 | - | 4, 5 | 721 | |||
| S62227.1 | - | 4, 5 | 721 | |||

Mnda
Genomic Map