siRNA Details
| Gene Symbol: 6330403A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081227.2 | NP_001074696.1 | 4 | 3234 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

6330403A02Rik| Gene Symbol: 6330403A02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081227.2 | NP_001074696.1 | 4 | 3234 | ||