siRNA Details
| Gene Symbol: Prdm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123362.1 | NP_001116834.1 | 5 | 1847 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdm12| Gene Symbol: Prdm12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123362.1 | NP_001116834.1 | 5 | 1847 | ||