siRNA Details
| Gene Symbol: Dfnb59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080711.2 | NP_001074180.1 | 5 | 902 | ||
| GenBank mRNA | DQ365828.1 | - | 5 | 902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dfnb59| Gene Symbol: Dfnb59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080711.2 | NP_001074180.1 | 5 | 902 | ||
| GenBank mRNA | DQ365828.1 | - | 5 | 902 | ||