siRNA Details
| Gene Symbol: Gm5149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132813.1 | - | 164 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5149| Gene Symbol: Gm5149 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK132813.1 | - | 164 | |||