siRNA Details
| Gene Symbol: Ube2u | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033773.4 | NP_001028945.2 | 8 | 934 | ||
| GenBank mRNA | AK133379.1 | - | 7 | 854 | ||
| BC150880.1 | - | 8 | 926 | 9087791 | ||
| BC150888.1 | - | 8 | 926 | 9087799 | ||
| BC172031.1 | - | 8 | 926 | 9054317 | ||

Ube2u
Genomic Map