siRNA Details
| Gene Symbol: Ccdc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033455.2 | NP_001028627.1 | 1, 2 | 272 | ||
| GenBank mRNA | AK133459.1 | - | 1, 2 | 273 | ||
| BC150930.1 | - | 1, 2 | 255 | 9087841 | ||
| BC151167.1 | - | 1, 2 | 255 | 9088056 | ||

Ccdc27
Genomic Map