siRNA Details
| Gene Symbol: Tmem240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101506.1 | NP_001094976.1 | 4 | 1158 | ||
| GenBank mRNA | AK147532.1 | - | 2 | 5290 | ||
| CJ257322.1 | - | 1 | 285 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem240| Gene Symbol: Tmem240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101506.1 | NP_001094976.1 | 4 | 1158 | ||
| GenBank mRNA | AK147532.1 | - | 2 | 5290 | ||
| CJ257322.1 | - | 1 | 285 | |||