siRNA Details
| Gene Symbol: 1700015F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200025.1 | NP_001186954.1 | 2 | 313 | ||
| GenBank mRNA | AK005993.1 | - | 2 | 262 | ||
| BC052364.1 | - | 3 | 658 | |||
| BC099562.1 | - | 2 | 272 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700015F17Rik| Gene Symbol: 1700015F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200025.1 | NP_001186954.1 | 2 | 313 | ||
| GenBank mRNA | AK005993.1 | - | 2 | 262 | ||
| BC052364.1 | - | 3 | 658 | |||
| BC099562.1 | - | 2 | 272 | |||