siRNA Details
Gene Symbol: 1700015F17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200025.1 | NP_001186954.1 | 4, 5 | 522 | ||
GenBank mRNA | AK005993.1 | - | 4, 5 | 472 | ||
BC052364.1 | - | 5, 6 | 867 | |||
BC099562.1 | - | 4, 5 | 481 |
