siRNA Details
| Gene Symbol: Gm1070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136050.1 | - | 2 | 1046 | ||
| BC147402.1 | - | 2 | 1025 | 9056454 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm1070| Gene Symbol: Gm1070 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK136050.1 | - | 2 | 1046 | ||
| BC147402.1 | - | 2 | 1025 | 9056454 | ||