siRNA Details
| Gene Symbol: Lpar5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163268.1 | NP_001156740.1 | 2 | 1239 | ||
| NM_001163269.1 | NP_001156741.1 | 2 | 1174 | |||
| GenBank mRNA | BC116702.1 | - | 2 | 1139 | 40104865 | |
| BC117528.1 | - | 2 | 1139 | 40104864 | ||
More Information
Set Membership: |
|

Lpar5
Genomic Map