siRNA Details
| Gene Symbol: Psen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019163.3 | NP_062036.2 | 2 | 187 | ||
| XM_006240321.2 | XP_006240383.1 | 4 | 457 | |||
| XM_006240322.2 | XP_006240384.1 | 4 | 710 | |||
| GenBank mRNA | BC070887.1 | - | 4 | 383 | 7098018 | |
| D82363.1 | - | 2 | 187 | |||
| D82578.1 | - | 2 | 187 | |||
| FQ231303.1 | - | 4 | 405 | |||

Psen1
Genomic Map