siRNA Details
| Gene Symbol: Sbk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200041.1 | NP_001186970.1 | 4 | 2545 | ||
| GenBank mRNA | AK138915.1 | - | 4 | 2547 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbk3| Gene Symbol: Sbk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200041.1 | NP_001186970.1 | 4 | 2545 | ||
| GenBank mRNA | AK138915.1 | - | 4 | 2547 | ||