siRNA Details
| Gene Symbol: Zfp882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166645.1 | NP_001160117.1 | 3 | 1726 | ||
| GenBank mRNA | BC080687.1 | - | 3 | 1667 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp882| Gene Symbol: Zfp882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166645.1 | NP_001160117.1 | 3 | 1726 | ||
| GenBank mRNA | BC080687.1 | - | 3 | 1667 | ||