siRNA Details
| Gene Symbol: Zfp882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166645.1 | NP_001160117.1 | 3 | 4438 | ||
| GenBank mRNA | AK048139.1 | - | 1 | 667 | ||
| AK140448.1 | - | 1 | 1084 | |||
| AK169264.1 | - | 1 | 649 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp882| Gene Symbol: Zfp882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166645.1 | NP_001160117.1 | 3 | 4438 | ||
| GenBank mRNA | AK048139.1 | - | 1 | 667 | ||
| AK140448.1 | - | 1 | 1084 | |||
| AK169264.1 | - | 1 | 649 | |||