siRNA Details
| Gene Symbol: Sptbn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019167.1 | NP_062040.1 | 2 | 533 | ||
| XM_008760094.1 | XP_008758316.1 | 2 | 262 | |||
| GenBank mRNA | AB001347.1 | - | 2 | 533 | ||
| AB008551.1 | - | 1 | 142 | |||
| AF225960.1 | - | 1 | 142 | |||

Sptbn2
Genomic Map