siRNA Details
| Gene Symbol: Adgrg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033468.3 | NP_001028640.2 | 12 | 2435 | ||
| NM_001145972.1 | NP_001139444.1 | 12 | 2443 | |||
| GenBank mRNA | AK028878.1 | - | 12 | 2436 | ||
| AK156147.1 | - | 12 | 2554 | |||
| AK156609.1 | - | 12 | 2554 | |||
More Information
Set Membership: |
|

Adgrg5
Genomic Map