siRNA Details
| Gene Symbol: Zkscan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177505.1 | NP_001170976.1 | 1 | 218 | ||
| GenBank mRNA | BC042569.1 | - | 2 | 499 | ||
| BC119591.1 | - | 1 | 218 | 40058904 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan7| Gene Symbol: Zkscan7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177505.1 | NP_001170976.1 | 1 | 218 | ||
| GenBank mRNA | BC042569.1 | - | 2 | 499 | ||
| BC119591.1 | - | 1 | 218 | 40058904 | ||