siRNA Details
Gene Symbol: Fbxw22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014395.2 | NP_001014417.1 | 3, 4 | 319 | ||
GenBank mRNA | BC084593.1 | - | 3, 4 | 323 | 30477093 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxw22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014395.2 | NP_001014417.1 | 3, 4 | 319 | ||
GenBank mRNA | BC084593.1 | - | 3, 4 | 323 | 30477093 |