siRNA Details
| Gene Symbol: Cpxcr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033471.3 | NP_001028643.2 | 3 | 847 | ||
| GenBank mRNA | AK133001.1 | - | 3 | 849 | ||
| BC147520.1 | - | 2 | 704 | 9056512 | ||
| BC147548.1 | - | 2 | 704 | 9056539 | ||

Cpxcr1
Genomic Map