siRNA Details
| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164683.1 | NP_001158155.1 | 1 | 68 | ||
| GenBank mRNA | AK012546.1 | - | 1 | 29 | ||
| AK154711.1 | - | 1 | 72 | |||
| BY288280.1 | - | 1 | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem29| Gene Symbol: Tmem29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164683.1 | NP_001158155.1 | 1 | 68 | ||
| GenBank mRNA | AK012546.1 | - | 1 | 29 | ||
| AK154711.1 | - | 1 | 72 | |||
| BY288280.1 | - | 1 | 68 | |||