siRNA Details
| Gene Symbol: Odf3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033473.2 | NP_001028645.1 | 4 | 1158 | ||
| GenBank mRNA | AK158063.1 | - | 4 | 1159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Odf3l2| Gene Symbol: Odf3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033473.2 | NP_001028645.1 | 4 | 1158 | ||
| GenBank mRNA | AK158063.1 | - | 4 | 1159 | ||