siRNA Details
| Gene Symbol: Odf3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033473.2 | NP_001028645.1 | 4 | 784 | ||
| GenBank mRNA | AK158063.1 | - | 4 | 785 | ||
| BC147356.1 | - | 4 | 772 | 9056405 | ||
| BC147357.1 | - | 4 | 772 | 9056398 | ||
| BC172043.1 | - | 3 | 664 | 9054329 | ||

Odf3l2
Genomic Map