siRNA Details
| Gene Symbol: Colq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009937.2 | NP_034067.2 | 15 | 1307 | ||
| GenBank mRNA | AK134212.1 | - | 15 | 1311 | ||
| BC107386.2 | - | 15 | 1241 | 40057706 | ||
| BC107387.1 | - | 14 | 1104 | 40057715 | ||

Colq
Genomic Map