siRNA Details
Gene Symbol: Zfp488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013777.2 | NP_001013799.2 | 2 | 2901 | ||
GenBank mRNA | BC089025.1 | - | 2 | 2901 | 30618054 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013777.2 | NP_001013799.2 | 2 | 2901 | ||
GenBank mRNA | BC089025.1 | - | 2 | 2901 | 30618054 |