siRNA Details
| Gene Symbol: Kctd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026135.1 | NP_080411.1 | 1 | 316 | ||
| GenBank mRNA | AK220381.1 | - | 1 | 290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd16| Gene Symbol: Kctd16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026135.1 | NP_080411.1 | 1 | 316 | ||
| GenBank mRNA | AK220381.1 | - | 1 | 290 | ||