siRNA Details
| Gene Symbol: Gpr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101516.1 | NP_001094986.1 | 1 | 1354 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr25| Gene Symbol: Gpr25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101516.1 | NP_001094986.1 | 1 | 1354 | ||
Set Membership: |
|