siRNA Details
| Gene Symbol: Tyms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019179.1 | NP_062052.1 | 1 | 94 | ||
| GenBank mRNA | BC126093.1 | - | 1 | 108 | 8362547 | |
| L12138.1 | - | 1 | 94 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tyms| Gene Symbol: Tyms | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019179.1 | NP_062052.1 | 1 | 94 | ||
| GenBank mRNA | BC126093.1 | - | 1 | 108 | 8362547 | |
| L12138.1 | - | 1 | 94 | |||