siRNA Details
Gene Symbol: Ankrd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081971.1 | NP_001075440.1 | 1 | 4257 | ||
GenBank mRNA | AK087787.1 | - | 1 | 2947 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ankrd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081971.1 | NP_001075440.1 | 1 | 4257 | ||
GenBank mRNA | AK087787.1 | - | 1 | 2947 |