siRNA Details
Gene Symbol: Pramel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085418.2 | NP_001078887.1 | 4 | 1384 | ||
GenBank mRNA | BC052827.1 | - | 4 | 1394 | 30033377 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pramel5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085418.2 | NP_001078887.1 | 4 | 1384 | ||
GenBank mRNA | BC052827.1 | - | 4 | 1394 | 30033377 |