siRNA Details
| Gene Symbol: Fam47e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033478.2 | NP_001028650.2 | 2 | 219 | ||
| NM_001170571.1 | NP_001164042.1 | 2 | 219 | |||
| GenBank mRNA | AA244788.1 | - | 2 | 200 | ||
| AK144479.1 | - | 2 | 219 | |||
| BE650192.1 | - | 3 | 225 | |||
| DV648863.1 | - | 2 | 279 | |||

Fam47e
Genomic Map