siRNA Details
| Gene Symbol: Scgb1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256073.1 | NP_001243002.1 | 2, 3 | 262 | ||
| GenBank mRNA | CK616146.1 | - | 2, 3 | 280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1b3| Gene Symbol: Scgb1b3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256073.1 | NP_001243002.1 | 2, 3 | 262 | ||
| GenBank mRNA | CK616146.1 | - | 2, 3 | 280 | ||