siRNA Details
| Gene Symbol: Gm5347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079931.2 | NP_001073400.1 | 3 | 1196 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5347| Gene Symbol: Gm5347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079931.2 | NP_001073400.1 | 3 | 1196 | ||