siRNA Details
Gene Symbol: Gm1527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033479.4 | NP_001028651.1 | 2, 3 | 294 | ||
GenBank mRNA | AK132999.1 | - | 2, 3 | 293 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm1527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033479.4 | NP_001028651.1 | 2, 3 | 294 | ||
GenBank mRNA | AK132999.1 | - | 2, 3 | 293 |