siRNA Details
| Gene Symbol: Gm5382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034100.1 | NP_001029272.1 | 1 | 348 | ||
| GenBank mRNA | BC100482.1 | - | 1 | 348 | 30918325 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5382| Gene Symbol: Gm5382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034100.1 | NP_001029272.1 | 1 | 348 | ||
| GenBank mRNA | BC100482.1 | - | 1 | 348 | 30918325 | |