siRNA Details
| Gene Symbol: Frmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190332.1 | NP_001177261.1 | 12 | 4005 | ||
| GenBank mRNA | AK032347.1 | - | 1 | 2336 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frmd7| Gene Symbol: Frmd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190332.1 | NP_001177261.1 | 12 | 4005 | ||
| GenBank mRNA | AK032347.1 | - | 1 | 2336 | ||