siRNA Details
| Gene Symbol: 4932443I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101519.1 | NP_001094989.1 | 4 | 651 | ||
| GenBank mRNA | AK038767.1 | - | 4 | 653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4932443I19Rik| Gene Symbol: 4932443I19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101519.1 | NP_001094989.1 | 4 | 651 | ||
| GenBank mRNA | AK038767.1 | - | 4 | 653 | ||