siRNA Details
| Gene Symbol: Olfr250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207552.2 | NP_997435.2 | 1 | 181 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr250| Gene Symbol: Olfr250 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207552.2 | NP_997435.2 | 1 | 181 | ||