siRNA Details
| Gene Symbol: Olfr1383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207574.1 | NP_997457.1 | 1 | 925 | ||
| GenBank mRNA | BC127965.1 | - | 1 | 929 | 40134985 | |
| Gene Symbol: Olfr1372-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK137189.1 | - | 2225 | |||
| BC055827.1 | - | 2 | 1286 | |||

Olfr1383
Genomic Map