siRNA Details
| Gene Symbol: Sepp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083911.1 | NP_001077380.1 | 2 | 221 | ||
| NM_019192.2 | NP_062065.2 | 2 | 149 | |||
| GenBank mRNA | BC059137.1 | - | 2 | 150 | ||
| BC072539.1 | - | 2 | 138 | 7104195 | ||
| CB699091.1 | - | 2 | 221 | |||
| CB745388.1 | - | 2 | 373 | |||
| D25221.1 | - | 2 | 130 | |||
| DN933732.1 | - | 2 | 469 | |||
| M63574.1 | - | 109 | ||||

Sepp1
Genomic Map