siRNA Details
| Gene Symbol: Olfr1082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207674.2 | NP_997557.2 | 2 | 947 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1082| Gene Symbol: Olfr1082 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207674.2 | NP_997557.2 | 2 | 947 | ||