siRNA Details
Gene Symbol: Gm5415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164286.1 | NP_001157758.1 | 1 | 390 | ||
GenBank mRNA | AK029494.1 | - | 1 | 391 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164286.1 | NP_001157758.1 | 1 | 390 | ||
GenBank mRNA | AK029494.1 | - | 1 | 391 |