siRNA Details
Gene Symbol: Gm9839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199956.1 | NP_001186885.1 | 1 | 849 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm9839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199956.1 | NP_001186885.1 | 1 | 849 |