siRNA Details
Gene Symbol: BC024978 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243888.1 | NP_001230817.1 | 7 | 1552 | ||
NM_001290511.1 | NP_001277440.1 | 7 | 1397 | |||
GenBank mRNA | AK144930.1 | - | 2 | 435 | ||
BC024978.1 | - | 6 | 853 |
