siRNA Details
| Gene Symbol: BC024978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243888.1 | NP_001230817.1 | 1 | 91 | ||
| NM_001290511.1 | NP_001277440.1 | 1 | 91 | |||
| NM_001290513.1 | NP_001277442.1 | 1 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC024978| Gene Symbol: BC024978 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243888.1 | NP_001230817.1 | 1 | 91 | ||
| NM_001290511.1 | NP_001277440.1 | 1 | 91 | |||
| NM_001290513.1 | NP_001277442.1 | 1 | 91 | |||