siRNA Details
| Gene Symbol: Gm11568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205030.1 | NP_001191959.1 | 1 | 1068 | ||
| GenBank mRNA | AK029194.1 | - | 1 | 1069 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11568| Gene Symbol: Gm11568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205030.1 | NP_001191959.1 | 1 | 1068 | ||
| GenBank mRNA | AK029194.1 | - | 1 | 1069 | ||